
Propyl benzoate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2315-68-6
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009370
Boiling Pt:231 °C (1013 hPa)
Melting Pt:–52 °C
Density:1,02 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:>100 °C
:164.2 g/mol
EINECS:219-020-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.35 g/l (25 °C)
- Melting Point:-52 °C
- Molar Mass:164.2 g/mol
- Boiling Point:231 °C (1013 hPa)
- Vapor Pressure:0.181 hPa (25 °C)
- Flash Point:100 °C
- Refractive Index:1.4981 (25 °C, 589 nm)
- Density:1.02 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together