Order Entry
Ireland
ContactUsLinkComponent
4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®
Irritant
4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
4-Hydroxybenzaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    13.8 g/l (30.5 °C)
  • Melting Point:
    116 °C
  • Molar Mass:
    122.12 g/mol
  • Boiling Point:
    (1013 hPa) (sublimed)
  • Flash Point:
    174 °C
  • Density:
    1.13 g/cm3 (20 °C)
  • Ignition Point:
    450 °C
  • Hill Formula:
    C7H6O2
  • Chemical Formula:
    4-(OH)C6H4CHO

Specification Test Results

Related Information

Frequently Bought Together