Order Entry
Ireland
ContactUsLinkComponent
2,4,6-Trimethoxybenzaldehyde 98%
2,4,6-Trimethoxybenzaldehyde 98%
Certificates
undefined
2,4,6-Trimethoxybenzaldehyde 98%

Specification Test Results

  • Appearance:
    Beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    118 to 120 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together