
p-Anisaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-11-5
:
Formula:C₈H₈O₂
MDL Number:MFCD00003385
Merck Index:14,663
Boiling Pt:247…249 °C
Melting Pt:0…2 °C
Density:1,121
Flash Pt:116 °C (241 °F)
:136.15 g/mol
EINECS:204-602-6
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Frequently Bought Together