4-Nitrobenzaldehyde for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:555-16-8
:
Formula:C₇H₅NO₃
MDL Number:MFCD00007346
Merck Index:12,06684
Boiling Pt:300 °C (1013 hPa)
Melting Pt:106 °C
Density:1,546 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:151.12 g/mol
Storage Temperature:Ambient
EINECS:209-084-5
Specification Test Results
- Solubility:2.34 g/l (25 °C)
- Melting Point:101 - 104 °C
- Molar Mass:151.12 g/mol
- Bulk Density:400 kg/m3
Specification Test Results
Frequently Bought Together