
Vanillin for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:121-33-5
:
Formula:C₈H₈O₃
MDL Number:MFCD00006942
:152.15 g/mol
Merck Index:13,09998
Boiling Pt:285 °C (1013 hPa)
Melting Pt:∼82 °C
Density:1,056 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:153 °C
EINECS:204-465-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:10 g/l (25 °C)
- Melting Point:82 °C
- Molar Mass:152.14 g/mol
- Bulk Density:600 kg/m3
- Boiling Point:154 °C (13 hPa)
- Vapor Pressure:0.0029 hPa (25 °C)
- Flash Point:153 °C
- Density:1.06 g/cm3 (20 °C)
- pH:4.3 (10 g/l, H2O, 20 °C)
- Ignition Point:400 °C
Specification Test Results
Frequently Bought Together