Benzaldehyde 99.5+%, AcroSeal®, pure, redistilled
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-52-7
:
Formula:C₆H₅CHO
MDL Number:MFCD00003299
:106.12 g/mol
Merck Index:13,01057
Boiling Pt:179 °C (1013 hPa)
Melting Pt:–26 °C
Density:1,043 g/cm³
Flash Pt:64 °C
Storage Temperature:Ambient
EINECS:202-860-4
UN:1990
ADR:9,III
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5440 to 1.5460 (20°C, 589 nm)
- GC:≥99.50 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO37836
SDS
AcroSeal Brochure VWR Final AO_AcroSeal_Product Lists_VWR_final
Product Literature
Frequently Bought Together