2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
: Merck
:3682-35-7
:
:C₁₈H₁₂N₆
:MFCD00006045
:312.33 g/mol
:M-Clarity™ quality level = MQ100
:Ambient
:222-965-9
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Iron(II) sulphate heptahydrate ≥98%
: Thermo Fisher Scientific