Organic reference standard, Bis(4-hydroxyphenyl) sulphone 500 µg/ml in methanol
PROOSL20255
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:80-09-1
- Description:Bis(4-hydroxyphenyl) sulphone 500 µg/ml in methanol
- Formula:C₁₂H₁₀O₄S
- Description:Bis(4-hydroxyphenyl) sulphone