2,4,6-Triallyloxy-1,3,5-triazine stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:101-37-1
:
Formula:C₁₂H₁₅N₃O₃
MDL Number:MFCD00006049
:249.27 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ200
EINECS:202-936-7
UN:3077
:GHS07
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:0.3 g/l (20 °C)
- Melting Point:26 - 27 °C
- Molar Mass:249.27 g/mol
- Boiling Point:149 - 150 °C (3 hPa) decomposes
- Vapor Pressure:1.3 hPa (100 °C)
- Flash Point:165 - 170 °C
- Density:1.000 g/cm3 (20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specification Test Results
Frequently Bought Together