2,4,6-Tris(2-pyridyl)-1,3,5-triazine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3682-35-7
:
Formula:C₁₈H₁₂N₆
MDL Number:MFCD00006045
:312.33 g/mol
Merck Index:14,9750
Melting Pt:248…252 °C
EINECS:222-965-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together