
2-Nitrophenylhydrazine, moistened 30% with water 97%
Catalog # ACRO128830100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Orange to orange-brown to dark red Powder
- Infrared spectrum:Conforms
- Water:30 to 35 %
- HPLC:≥96.0 %
- Elemental Analysis:14.36 to 15.10 % H
Specification Test Results
About this item
:
:153.14 g/mol
Melting Pt:89…94 °C
Storage Temperature:Refrigerator
EINECS:221-222-6
UN:1325
ADR:4.1,II
:GHS07
CAS Number:3034-19-3
Formula:C₆H₇N₃O₂