1-Bromo-3-fluorobenzene
Catalog # MOLE12409749-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:175 g/mol
Boiling Pt:148 °C (1013 hPa)
Melting Pt:–8 °C
Density:1,7081 g/cm³ (20 °C)
Flash Pt:46 °C
Storage Temperature:Ambient
EINECS:214-023-0
UN:1993
ADR:3,III
CAS Number:1073-06-9
Formula:C₆H₄BrF
MDL Number:MFCD00000326