Order Entry
Ireland
ContactUsLinkComponent
 
1-Bromo-3-fluorobenzene 99%
 
undefined
1-Bromo-3-fluorobenzene 99%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5260 to 1.5280 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together