1-Bromo-2-fluorobenzene
Catalog # MOLEM59737074
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:175 g/mol
Boiling Pt:155 °C (1013 hPa)
Density:1,61 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-018-3
UN:1993
ADR:3,III
CAS Number:1072-85-1
Formula:C₆H₄BrF
MDL Number:MFCD00000282