
4-Bromo-3-chloroaniline
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:21402-26-6
:
Formula:C₆H₅BrClN
MDL Number:MFCD00041311
:206.47 g/mol
Boiling Pt:264 °C (1013 hPa)
Melting Pt:67…68 °C
EINECS:244-370-3
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together