Order Entry
Ireland
ContactUsLinkComponent
Anthracene, extra pure
Environment
Anthracene, extra pure
Catalog # 122892W
Supplier:  VWR Chemicals
CAS Number:  
undefined
Environment
Anthracene, extra pure
Catalog # 122892W
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :178.23 g/mol
Merck Index:13,00687
Boiling Pt:340 °C (1013 hPa)
Melting Pt:111…112 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-371-1
UN:3077
ADR:9,III
 :GHS09
CAS Number:120-12-7
Formula:C₁₄H₁₀
MDL Number:MFCD00001240