Order Entry
Ireland
ContactUsLinkComponent
Disposable earplugs, E-A-R™ Classic™ Soft
  111-1104
 :  3M
undefined
Disposable earplugs, E-A-R™ Classic™ Soft
  111-1104
 :  3M
  • Colour:
    Yellow
  • Description:
    Soft earplugs, without cord, 200 pairs per box
  • Size:
    U*
  • Pk:
    200 PAIR
These earplugs feature a unique PU-foam combination which warms to the body temperature and becomes softer, thus allowing the earplug to mould to the individuals ear canal shape.
  • Efficient seal due to the moulding of the earplug to the ear canal
  • Textured surface provides greater friction
  • Improves grip and helps maintain an effective seal
Resistant to moisture for hot and humid working environments.
SNR: 36 dB
 : EN 352-2
 : * One size fits all.