

2-Butyne 98%
:503-17-3
:
:C₄H₆
:MFCD00009275
:27 °C (1013 hPa)
:–32 °C
:0,69 g/cm³
:–25 °C
:Refrigerator
:207-962-2
:3,I
:54.09 g/mol
:1144
:GHS07
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3920 to 1.3940 (20°C, 589 nm)
- GC:≥97.5 %
Frequently Bought Together