


1-Octyne 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:629-05-0
:
Formula:C₈H₁₄
MDL Number:MFCD00009546
Boiling Pt:127…128 °C (760 mmHg)
Melting Pt:–80 °C
Density:0,74 g/cm³
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:211-069-3
ADR:3,II
:110.2 g/mol
UN:3295
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4150 to 1.4170 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together