1-Chloro-2-methylpropane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:513-36-0
:
Formula:C₄H₉Cl
MDL Number:MFCD00000954
:92.57 g/mol
Merck Index:13,05155
Boiling Pt:68 °C (1013 hPa)
Melting Pt:–131 °C
Density:0,8829 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-157-9
UN:1127
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–17 °C
:GHS02
Specification Test Results
- Solubility:28 mg/l (20 °C)
- Melting Point:-131 °C
- Molar Mass:92.56 g/mol
- Boiling Point:69 °C (1013 hPa)
- Vapor Pressure:161 hPa (20 °C)
- Flash Point:-17 °C
- Refractive Index:1.3975 (20 °C, 589 nm)
- Explosion Limit:2.0 - 8.8 %(V)
- Density:0.88 g/cm3 (20 °C)
- Ignition Point:395 °C
- Synonyms:Isobutyl chloride
- Hill Formula:C4H9Cl
- Chemical Formula:(CH3)2CHCH2Cl
Specification Test Results
Frequently Bought Together