1-Octene, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:111-66-0
:
Formula:C₈H₁₆
MDL Number:MFCD00009548
:112.22 g/mol
Merck Index:13,01770
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–102 °C
Density:0,7152 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-893-7
UN:3295
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:8 °C
Frequently Bought Together