



2,2,4-Trimethylpentane ≥99.5%, Environmental Grade
Catalog # 41904.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:114.23 g/mol
Merck Index:14,5193
Boiling Pt:98…99 °C
Melting Pt:–107 °C
Density:0,692
Flash Pt:–7 °C (19 °F)
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:540-84-1
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943