n-Octane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:111-65-9
:
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556
:114.23 g/mol
Merck Index:13,06782
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–57 °C
Density:0,703 g/cm³ (20 °C)
Flash Pt:8 °C
EINECS:203-892-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Boiling point:125 - 127 °C
- Density (20/4):0.700 - 0.710
- Evaporation residue:Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Glass bottle for liquids DIN 32 (ZPACK13415030) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
28746348.pdf spec_VWRC26025.238 spec_VWRC26025.238 spec_VWRC26025.238 spec_VWRC26025.466
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together