Order Entry
Ireland
ContactUsLinkComponent
1-Ethoxy-2,2,2-trifluoroethanol 90%
1-Ethoxy-2,2,2-trifluoroethanol 90%
Certificates
undefined
1-Ethoxy-2,2,2-trifluoroethanol 90%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3410 to 1.3440 (20°C, 589 nm)
  • GC:
    ≥88 %

Specification Test Results

Related Information

Frequently Bought Together