Hexane (mixture of isomers), Environmental Grade for environmental analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Hexanes, Isohexane
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:65…70 °C
Melting Pt:–95 °C
Density:0,67
Flash Pt:<–23 °C (–9 °F)
EINECS:295-570-2
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together