
2-Butanethiol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:513-53-1
:
Formula:C₄H₁₀S
MDL Number:MFCD00004865
:90.19 g/mol
Merck Index:12,01612
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–165 °C
Density:0,829 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-165-2
UN:2347
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–23 °C
:GHS02
Specification Test Results
- Solubility:1.32 g/l (20 °C)
- Melting Point:-165 °C
- Molar Mass:90.18 g/mol
- Boiling Point:82 - 88 °C (1013 hPa)
- Vapor Pressure:108 hPa (25 °C)
- Flash Point:-23 °C
- Explosion Limit:1.7 - 9.6 %(V)
- Density:0.83 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together