Order Entry
Ireland
Orders LinkContactUsLinkComponent
1-Hexanol 98%, pure

1-Hexanol 98%, pure

Flammable
Irritant
1-Hexanol 98%, pure
Certificates
1-Hexanol 98%, pure

5 Options Available Below

Product Details & Documents
Synonyms:  Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
  • CAS Number:111-27-3
  • Formula:H₃C(CH₂)₄CH₂OH
  • MDL Number:MFCD00002982
  • Molecular Weight:102.18 g/mol
  • Merck Index:13,04715
  • Boiling Pt:156…157 °C (760 mmHg)
  • Melting Pt:–52 °C
  • Density:0,8136 g/cm³
  • Flash Pt:59 °C
  • Storage Temperature:Ambient
  • EINECS:203-852-3
  • UN:2282
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4170 to 1.4190 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥97.5 %
  • Color scale:
    ≤10 APHA
  • Acid value:
    ≤0.02 mg KOH/g
  • Ester value:
    ≤0.3 mg KOH/g
  • Iodine value:
    ≤0.1 g I2/100 g
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 30,50 €
    30,50 €
    1 * 100 mL
       30,50 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 59,20 €
    59,20 €
    1 * 1 L
       59,20 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 117,00 €
    117,00 €
    1 * 2,5 L
       117,00 €

  • Plastic drum
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 410,00 €
    410,00 €
    1 * 10 L
       410,00 €

  • Metal drum
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 778,00 €
    778,00 €
    1 * 25 L
       778,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.