

1-Decanol
Catalog # PROOCHE172
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
:158.28 g/mol
Merck Index:13,02875
Boiling Pt:230 °C (1013 hPa)
Melting Pt:7 °C
Density:0,82634 g/cm³ (20 °C)
Flash Pt:82 °C
Storage Temperature:Ambient
EINECS:203-956-9
UN:3082
ADR:9,III
:GHS07
:GHS09
CAS Number:112-30-1
Formula:H₃C(CH₂)₈CH₂OH
MDL Number:MFCD00004747