


Formaldehyde dimethyl acetal for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-87-5
:
Formula:CH₂(OCH₃)₂
MDL Number:MFCD00008495
:76.1 g/mol
Merck Index:12,06093
Boiling Pt:42 °C (1013 hPa)
Melting Pt:–105 °C
Density:0,821 g/cm³ (20 °C)
EINECS:203-714-2
UN:1234
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–18 °C
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-105 °C
- Molar Mass:76.11 g/mol
- Boiling Point:42.3 °C (1013 hPa)
- Vapor Pressure:440 hPa (20 °C)
- Flash Point:-18 °C
- Refractive Index:1.3541 (20 °C, 589 nm)
- Explosion Limit:1.6 - 17.6 %(V)
- Density:0.86 g/cm3 (20 °C)
- Ignition Point:235 °C
Specification Test Results
Frequently Bought Together