Order Entry
Ireland
Orders LinkContactUsLinkComponent
Diethyl ether

Diethyl ether

Glass bottle, 2,5 L
Catalog # ROTHT900.1
Flammable
Irritant
Marketsource
Supplier:   Roth Carl
Diethyl ether
Catalog # ROTHT900.1
Supplier:  Roth Carl
Marketsource
CAS Number:  60-29-7
Diethyl ether
Diethyl ether

Glass bottle, 2,5 L

CAS Number:  60-29-7
Price
146,26 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0,7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • UN:1155
  • ADR:3,I
  • REACH:01-2119535785-29
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.