Isopropylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-31-0
:
Formula:C₃H₉N
MDL Number:MFCD00008082
:59.11 g/mol
Merck Index:13,05229
Boiling Pt:32,4 °C (1013 hPa)
Melting Pt:–101 °C
Density:0,69 g/cm³ (20 °C)
EINECS:200-860-9
UN:1221
ADR:3 (8),I
:M-Clarity™ quality level = MQ200
Flash Pt:–27 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Melting Point:-101 °C
- Molar Mass:59.11 g/mol
- Boiling Point:31 - 33 °C (1013 hPa)
- Vapor Pressure:632 hPa (20 °C)
- Flash Point:-37 °C
- Refractive Index:1.3746 (20 °C, 589 nm)
- Explosion Limit:2 - 10.4 %(V)
- Density:0.69 g/cm3 (20 °C)
- pH:13 (700 g/l, H2O, 20 °C)
- Ignition Point:330 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Related Information
Frequently Bought Together