Order Entry
Ireland
ContactUsLinkComponent
Dodecylamine for synthesis, Sigma-Aldrich®
Corrosive
Environment
Irritant
Dodecylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Environment
Irritant
Dodecylamine for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    25 - 28 °C
  • Molar Mass:
    185.35 g/mol
  • Boiling Point:
    247 - 249 °C (1013 hPa)
  • Vapor Pressure:
     - 2 hPa (20 °C)
  • Flash Point:
    110 °C
  • Refractive Index:
    1.4377 (30 °C)
  • Density:
    0.80 g/cm3 (20 °C)
  • Synonyms:
    Laurylamine, 1-Aminododecan
  • Hill Formula:
    C12H27N
  • Chemical Formula:
    CH3(CH2)11NH2

Specification Test Results

Related Information

Frequently Bought Together