2-Iodoethanol stabilised for synthesis, Sigma-Aldrich®
8.41353.9999
: Merck
:
- Pk:1
- Seq:0001
- Environmentally Preferable:
- Molar Mass:171.97 g/mol
- Boiling Point:172 - 175 °C (1013 hPa)
- Flash Point:65 °C
- Density:2.202 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
:
:171.97 g/mol
:85
:210-861-6
:2810
:GHS06
:Refrigerator
:624-76-0
:C₂H₅IO
:MFCD00002830
:2.202 g/cm³ (20 °C)
:6.1 II
:M-Clarity™ quality level = MQ200