Order Entry
Ireland
ContactUsLinkComponent
Trifluoroacetic acid 99%, extra pure
Irritant
Corrosive
Trifluoroacetic acid 99%, extra pure
Certificates
undefined
Irritant
Corrosive
Trifluoroacetic acid 99%, extra pure
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 % (Coulometric)
  • Color scale:
    ≤20 APHA
  • Titration with NaOH:
    99.0 to 101.0 %

Specification Test Results

Related Information

Frequently Bought Together