Bis(2-chloroethoxy)methane
Catalog # PROOCILDLM19450.1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
:
Boiling Pt:218,1 °C (1013 hPa)
Melting Pt:–32,8 °C
Density:1,2406 g/cm³ (11 °C)
:173.04 g/mol
EINECS:203-920-2
CAS Number:111-91-1
Formula:C₅H₁₀Cl₂O₂