Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Chloro-5-iodopentane
1-Chloro-5-iodopentane ≥97%
1-Chloro-5-iodopentane ≥97%
L04469.09
:
Thermo Fisher Scientific
:
1-Chloro-5-iodopentane ≥97%
L04469.09
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
60274-60-4
:
:
C₅H₁₀ClI
:
232.49 g/mol
:
75…77 °C (4 mmHg)
:
1,663
:
>110 °C (230 °F)
:
262-137-4
:
GHS07
:
Ambient
:
MFCD00039416