Order Entry
Ireland
ContactUsLinkComponent
Pesticide reference standard, 1,2,3-Trichloropropane 1000 µg/ml in methanol, CIL
Pesticide reference standard, 1,2,3-Trichloropropane 1000 µg/ml in methanol, CIL
Catalog # PROOCILULM69111.2
CAS Number:  
Pesticide reference standard, 1,2,3-Trichloropropane 1000 µg/ml in methanol, CIL
Catalog # PROOCILULM69111.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    96-18-4
  • Description:
    1,2,3-Trichloropropane 1,000 µg/ml in methanol
  • Formula:
    C₃H₅Cl₃
  • Description:
    1,2,3-Trichloropropane

Specifications

About this item