Order Entry
Ireland
ContactUsLinkComponent
Caprylic acid for synthesis, Sigma-Aldrich®
Corrosive
Caprylic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Caprylic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    0.68 g/l (20 °C)
  • Melting Point:
    16 °C
  • Molar Mass:
    144.21 g/mol
  • Boiling Point:
    238 °C (1013 hPa)
  • Vapor Pressure:
    0.05 hPa (20 °C)
  • Flash Point:
    109.9 °C
  • Explosion Limit:
    1 %(V)
  • Density:
    0.91 g/cm3 (20 °C)
  • pH:
    4 (0.2 g/l, H2O, 20 °C)
  • Ignition Point:
    300 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together