
Caprylic acid Ph. Eur., SAFC®
Catalog # 1.00193.2500
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:144.21 g/mol
Merck Index:13,01771
Boiling Pt:239,7 °C (1013 hPa)
Melting Pt:16 °C
Density:0,9066 g/cm³ (20 °C)
Flash Pt:130 °C
Storage Temperature:Ambient
EINECS:204-677-5
UN:3265
ADR:8,III
:GHS05
CAS Number:124-07-2
Formula:C₈H₁₆O₂
MDL Number:MFCD00004429