Order Entry
Ireland
ContactUsLinkComponent
 
 
 
Ethyl acetate, Uvasol® for spectroscopy, Supelco®
 :  Merck
    
 
undefined
 
 
Ethyl acetate, Uvasol® for spectroscopy, Supelco®
 :  Merck
    

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Solubility:
    85.3 g/L (20 °C)
  • Melting Point:
    -83 °C
  • Molar Mass:
    88.11 g/mol
  • Boiling Point:
    77 °C (1013 hPa)
  • Vapor Pressure:
    97 hPa (20 °C)
  • Flash Point:
    -4 °C
  • Refractive Index:
    1.37
  • Explosion Limit:
    2.1 - 11.5%(V)
  • Density:
    0.90 g/cm³ (20 °C)
  • pH:
    (H₂O) no data available
  • Ignition Point:
    460 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

 

Frequently Bought Together