

Acetonylacetone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-13-4
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00008792
:114.14 g/mol
Merck Index:13,00072
Boiling Pt:191 °C (1013 hPa)
Melting Pt:–5,4 °C
Density:0,971 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:203-738-3
:GHS08
:GHS07
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-5.4 °C
- Molar Mass:114.14 g/mol
- Boiling Point:85 - 87 °C (26.6 hPa)
- Vapor Pressure:0.6 hPa (20 °C)
- Flash Point:79 °C
- Refractive Index:1.423 (20 °C, 589 nm)
- Explosion Limit:1.5 %(V)
- Density:0.974 g/cm3 (20 °C)
- pH:6.1 (10 g/l, H2O, 20 °C)
- Ignition Point:400 °C
Specification Test Results
Frequently Bought Together