Order Entry
Ireland
ContactUsLinkComponent
Acetone, Selectipur® for the electronics industry
Flammable
Irritant
Acetone, Selectipur® for the electronics industry
Catalog # 55954791.
CAS Number:  
undefined
Flammable
Irritant
Acetone, Selectipur® for the electronics industry
Catalog # 55954791.
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    146 kg
  • Seq:
    4595

Specifications

About this item

 :
 :58.08 g/mol
Merck Index:13,00067
Boiling Pt:56,2 °C (1013 hPa)
Melting Pt:–95,4 °C
Density:0,792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
 :GHS02
 :GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765