Methyl ethyl ketone, EMPLURA®, Supelco®
: Merck
:78-93-3
:
:C₂H₅COCH₃
:MFCD00011648
:13,06097
:79,6 °C (1013 hPa)
:–86 °C
:0,8054 g/cm³ (20 °C)
:–4 °C
:201-159-0
:1193
:3,II
:M-Clarity™ quality level = MQ200
:72.11 g/mol
:01-2119457290-43
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Frequently Bought Together