

4-Heptanone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-19-3
:
Formula:C₇H₁₄O
MDL Number:MFCD00009403
:114.19 g/mol
Merck Index:13,03379
Boiling Pt:145 °C (760 mmHg)
Melting Pt:–33 °C
Density:0,81 g/cm³
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:204-608-9
UN:2710
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4060 to 1.4080 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together