
2-Undecanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-12-9
:
Formula:C₁₁H₂₂O
MDL Number:MFCD00009583
Merck Index:12,06178
Boiling Pt:231 °C (1013 hPa)
Melting Pt:∼12 °C
Density:0,83 g/cm³ (20 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:203-937-5
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ200
:170.3 g/mol
:GHS09
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:11 - 13 °C
- Molar Mass:170.29 g/mol
- Boiling Point:228 °C (1013 mbar)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:96 °C
- Refractive Index:1.428 (20 °C, 589 nm)
- Density:0.83 g/cm3 (20 °C)
- Synonyms:Methyl nonyl ketone
- Hill Formula:C11H22O
- Chemical Formula:CH3(CH2)8COCH3
Specification Test Results
Frequently Bought Together