
2-Undecanone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-12-9
:
Formula:C₁₁H₂₂O
MDL Number:MFCD00009583
Merck Index:12,06178
Boiling Pt:231…232 °C (760 mmHg)
Melting Pt:11…13 °C
Density:0,824 g/cm³
Flash Pt:88 °C
UN:3082
ADR:9,III
:170.3 g/mol
EINECS:203-937-5
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4280 to 1.4310 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together