

2-Heptanone, Supelco®
Catalog # 02476-1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:114.19 g/mol
Merck Index:13,04683
Boiling Pt:150 °C (1013 hPa)
Melting Pt:–35 °C
Density:0,8111 g/cm³ (20 °C)
Flash Pt:43 °C
EINECS:203-767-1
UN:1110
ADR:3,III
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:110-43-0
Formula:C₇H₁₄O
MDL Number:MFCD00009513