Order Entry
Ireland
ContactUsLinkComponent
 
 
4-Methoxybenzamide 97%
 
undefined
 
4-Methoxybenzamide 97%

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    164 to 167 °C
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together