Order Entry
Ireland
ContactUsLinkComponent
 
 
4-(Bromomethyl)benzenesulphonyl chloride 95%
 
undefined
 
4-(Bromomethyl)benzenesulphonyl chloride 95%

 

  • Appearance:
    White to yellow to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    72 to 77 °C
  • Titration Argentometric:
    ≥94 %

 

 

Frequently Bought Together